Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000456.2 | NP_000447.2 | 5, 6 | 564 | ||
NM_001032386.1 | NP_001027558.1 | 4, 5 | 438 | |||
NM_001032387.1 | NP_001027559.1 | 3, 4 | 329 | |||
XM_005269112.1 | XP_005269169.1 | 4, 5 | 420 | |||
XM_017019905.1 | XP_016875394.1 | 6, 7 | 1110 | |||
XM_017019906.1 | XP_016875395.1 | 4, 5 | 398 | |||
XM_017019907.1 | XP_016875396.1 | 5, 6 | 1179 | |||
XM_017019908.1 | XP_016875397.1 | 3, 4 | 339 | |||
GenBank mRNA | AK312763.1 | - | 3, 4 | 303 | ||
AU121557.1 | - | 4, 5 | 410 | |||
AU126450.1 | - | 3, 4 | 329 | |||
BC065193.1 | - | 3, 4 | 360 | 5227360 | ||
HM013717.1 | - | 2, 3 | 247 | |||
L31573.1 | - | 2, 3 | 970 |
Gene Symbol: RAB11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647356.1 | - | 1630 |