Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297438.1 | NP_001284367.1 | 3 | 552 | ||
NM_014231.4 | NP_055046.1 | 3 | 552 | |||
NM_016830.3 | NP_058439.1 | 3 | 552 | |||
NM_199245.2 | NP_954740.1 | 3 | 552 | |||
NR_123717.1 | 2 | 425 | ||||
GenBank mRNA | AF060538.1 | - | 3 | 314 | ||
AK289498.1 | - | 3 | 323 | |||
AK296294.1 | - | 3 | 330 | |||
AK303137.1 | - | 3 | 330 | |||
BC023286.1 | - | 3 | 318 | 4943992 | ||
BE246448.1 | - | 3 | 330 | |||
BE797045.1 | - | 2 | 140 | |||
BI838752.1 | - | 3 | 347 | |||
BQ717992.1 | - | 3 | 311 | |||
CR542214.1 | - | 2 | 184 | |||
CR542231.1 | - | 2 | 184 | |||
CR749485.1 | - | 3 | 553 | |||
CR989458.1 | - | 3 | 306 | |||
Z48924.1 | - | 2 | 184 |
Gene Symbol: TAPBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098035.1 | - | 9 | 1769 |