Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHLDA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003311.3 | NP_003302.1 | 1 | 344 | ||
GenBank mRNA | AF001294.1 | - | 1 | 344 | ||
AF019953.1 | - | 1 | 305 | |||
AF035444.1 | - | 1 | 344 | |||
AK223027.1 | - | 1 | 344 | |||
BC005034.1 | - | 1 | 334 | 3954346 | ||
CR407664.1 | - | 1 | 288 |