Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TTC3 SIRNA Assays ›
Gene Symbol:
TTC3
Gene Name:
tetratricopeptide repeat domain 3
Gene Aliases:
DCRR1, RNF105, TPRDIII
Chromosome Location:
Chr.21: 37073184 - 37203118 on Build GRCh38
UniGene ID:
Hs.368214
Species:
Human
siRNA ID
HSS144356
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TTC3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001894.1
NP_001001894.1
35, 36
4729
NM_001320703.1
NP_001307632.1
36, 37
4989
NM_001320704.1
NP_001307633.1
36, 37
4979
NM_003316.3
NP_003307.3
35, 36
6020
XM_005261048.1
XP_005261105.1
37, 38
4889
XM_005261049.1
XP_005261106.1
37, 38
4812
XM_005261050.4
XP_005261107.1
37, 38
7774
XM_005261052.1
XP_005261109.1
36, 37
4764
XM_005261054.3
XP_005261111.1
36, 37
7718
XM_005261055.1
XP_005261112.1
27, 28
3944
XM_005261056.2
XP_005261113.1
27, 28
4023
XM_011529739.2
XP_011528041.1
29, 30
4095
XM_011529740.2
XP_011528042.1
33, 34
4549
XM_017028458.1
XP_016883947.1
37, 38
4828
XM_017028459.1
XP_016883948.1
36, 37
4773
XM_017028460.1
XP_016883949.1
35, 36
4724
XM_017028461.1
XP_016883950.1
36, 37
4835
XM_017028462.1
XP_016883951.1
36, 37
4758
XM_017028463.1
XP_016883952.1
35, 36
4787
XM_017028464.1
XP_016883953.1
27, 28
4063
XM_017028465.1
XP_016883954.1
33, 34
6174
XM_017028466.1
XP_016883955.1
26, 27
3985
XM_017028467.1
XP_016883956.1
26, 27
3890
GenBank mRNA
AY523634.1
-
8, 9
1798
BC137345.1
-
35, 36
5159
9021343
BU508034.1
-
3, 4
493
CR936616.1
-
20, 21
3325
D83077.1
-
35, 36
6020
D83327.1
-
32, 33
4372
D84294.1
-
34, 35
6093
D84295.1
-
33, 34
6007
D84296.1
-
25, 26
4431
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
ubiquitin-dependent protein catabolic process
protein K48-linked ubiquitination
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0