Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003327.3 | NP_003318.1 | 5 | 527 | ||
XM_011542074.2 | XP_011540376.1 | 3 | 881 | |||
XM_011542075.2 | XP_011540377.1 | 5 | 994 | |||
XM_011542076.2 | XP_011540378.1 | 3 | 881 | |||
XM_011542077.2 | XP_011540379.1 | 3 | 240 | |||
XM_017002231.1 | XP_016857720.1 | 6 | 1872 | |||
XM_017002232.1 | XP_016857721.1 | 6 | 1872 | |||
GenBank mRNA | BC105070.1 | - | 5 | 527 | 8144073 | |
BC105072.1 | - | 5 | 527 | 8144075 | ||
S76792.1 | - | 5 | 477 |