Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003356.3 | NP_003347.1 | 4 | 785 | ||
NM_022803.2 | NP_073714.1 | 4 | 785 | |||
GenBank mRNA | AF001787.1 | - | 4 | 613 | ||
AF011449.1 | - | 3 | 528 | |||
AK092685.1 | - | 4 | 432 | |||
AK290630.1 | - | 4 | 633 | |||
AK291889.1 | - | 4 | 599 | |||
KU178372.1 | - | 3 | 430 | |||
U82818.1 | - | 4 | 583 | |||
U84763.1 | - | 4 | 583 |