Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003367.3 | NP_003358.1 | 6 | 740 | ||
NM_207291.2 | NP_997174.1 | 5 | 539 | |||
XM_005259197.3 | XP_005259254.1 | 6 | 639 | |||
XM_011527260.2 | XP_011525562.1 | 6 | 639 | |||
XM_011527261.2 | XP_011525563.1 | 6 | 639 | |||
XM_017027199.1 | XP_016882688.1 | 5 | 438 | |||
GenBank mRNA | AK297022.1 | - | 6 | 610 | ||
BC049821.1 | - | 6 | 653 | 5483523 | ||
CR536504.1 | - | 6 | 616 | |||
M77476.1 | - | 3 | 340 | |||
S50537.1 | - | 2 | 280 | |||
X90824.1 | - | 4 | 479 | |||
X90825.1 | - | 5 | 524 | |||
X90826.1 | - | 6 | 622 |