Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321150.1 | NP_001308079.1 | 7 | 574 | ||
NM_003367.3 | NP_003358.1 | 9 | 967 | |||
NM_207291.2 | NP_997174.1 | 8 | 766 | |||
XM_005259197.3 | XP_005259254.1 | 9 | 866 | |||
XM_017027199.1 | XP_016882688.1 | 8 | 665 | |||
GenBank mRNA | AK297022.1 | - | 9 | 837 | ||
AY147880.1 | - | 7 | 450 | |||
BC049821.1 | - | 9 | 880 | 5483523 | ||
CR536504.1 | - | 9 | 843 | |||
M77476.1 | - | 6 | 567 | |||
S50537.1 | - | 5 | 507 | |||
X90825.1 | - | 8 | 751 | |||
X90826.1 | - | 9 | 849 |