Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SF1 SIRNA Assays ›
Gene Symbol:
SF1
Gene Name:
splicing factor 1
Gene Aliases:
BBP, D11S636, MBBP, ZCCHC25, ZFM1, ZNF162
Chromosome Location:
Chr.11: 64764604 - 64779043 on Build GRCh38
UniGene ID:
Hs.502829
Species:
Human
siRNA ID
HSS144483
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178030.1
NP_001171501.1
2
543
NM_001178031.1
NP_001171502.1
2
325
NM_004630.3
NP_004621.2
2
546
NM_201995.2
NP_973724.1
2
546
NM_201997.2
NP_973726.2
2
546
NM_201998.2
NP_973727.1
2
546
NR_033649.1
2
546
NR_033650.1
2
546
XM_011545244.1
XP_011543546.1
2
218
XM_011545245.1
XP_011543547.1
2
218
XM_011545246.1
XP_011543548.1
2
218
XM_011545247.1
XP_011543549.1
2
218
XM_011545248.1
XP_011543550.1
2
218
XM_017018244.1
XP_016873733.1
2
218
XM_017018245.1
XP_016873734.1
2
299
XM_017018246.1
XP_016873735.1
2
218
XM_017018247.1
XP_016873736.1
2
261
XM_017018248.1
XP_016873737.1
2
261
XM_017018249.1
XP_016873738.1
2
745
XM_017018250.1
XP_016873739.1
2
471
XM_017018251.1
XP_016873740.1
2
326
GenBank mRNA
AK293753.1
-
2
325
AK296237.1
-
2
244
AK301803.1
-
2
471
BC008080.2
-
2
242
2961666
BC008724.2
-
2
242
2961666
BC020217.1
-
2
445
4869292
BC034451.1
-
2
471
BC038446.1
-
2
566
5493676
BX095574.1
-
2
211
D26120.1
-
2
476
D26121.1
-
2
476
DC368015.1
-
2
502
DC370979.1
-
2
546
L49345.1
-
2
432
L49380.1
-
2
432
Y08765.1
-
2
136
Y08766.1
-
2
388
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
spliceosomal complex assembly
mRNA 3'-splice site recognition
mRNA splicing, via spliceosome
transcription, DNA-templated
regulation of transcription, DNA-templated
male sex determination
Leydig cell differentiation
negative regulation of smooth muscle cell proliferation
regulation of steroid biosynthetic process
negative regulation of nucleic acid-templated transcription
transcription corepressor activity
RNA binding
protein binding
zinc ion binding
identical protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0