Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003534.2 | NP_003525.1 | 1 | 201 | ||
GenBank mRNA | BC012185.1 | - | 1 | 632 | ||
BC079835.1 | - | 1 | 206 |