Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130831.1 | NP_001124303.1 | 5 | 458 | ||
NM_003644.2 | NP_003635.2 | 1 | 207 | |||
NM_201432.1 | NP_958836.1 | 5 | 615 | |||
NM_201433.1 | NP_958839.1 | 5 | 645 | |||
XM_005256831.3 | XP_005256888.1 | 6 | 556 | |||
XM_005256832.3 | XP_005256889.1 | 5 | 426 | |||
XM_005256833.3 | XP_005256890.1 | 4 | 533 | |||
XM_005256834.3 | XP_005256891.1 | 4 | 359 | |||
XM_005256835.3 | XP_005256892.1 | 4 | 303 | |||
XM_011524044.2 | XP_011522346.1 | 5 | 450 | |||
XM_017025239.1 | XP_016880728.1 | 5 | 479 | |||
XM_017025240.1 | XP_016880729.1 | 4 | 262 | |||
XM_017025241.1 | XP_016880730.1 | 4 | 238 | |||
XM_017025242.1 | XP_016880731.1 | 4 | 412 | |||
GenBank mRNA | AB007854.1 | - | 5 | 413 | ||
AB209030.1 | - | 4 | 302 | |||
AJ224876.1 | - | 1 | 230 | |||
AK225165.1 | - | 5 | 437 | |||
AK292987.1 | - | 5 | 431 | |||
AK293834.1 | - | 1 | 219 | |||
AK294829.1 | - | 4 | 406 | |||
AK299574.1 | - | 5 | 437 | |||
AK307987.1 | - | 6 | 467 | |||
AK309722.1 | - | 5 | 466 | |||
AK315162.1 | - | 1 | 220 | |||
AY327406.1 | - | 5 | 645 | |||
BC001152.1 | - | 5 | 438 | 3353809 | ||
BC006454.1 | - | 5 | 615 | 3547000 | ||
BP202727.1 | - | 5 | 458 | |||
BT006891.1 | - | 4 | 292 |