Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320099.1 | NP_001307028.1 | 3 | 803 | ||
NM_003805.4 | NP_003796.1 | 3 | 593 | |||
GenBank mRNA | AB451280.1 | - | 2 | 485 | ||
AB451412.1 | - | 2 | 485 | |||
AK314861.1 | - | 3 | 573 | |||
BC017042.1 | - | 3 | 565 | 3849848 | ||
BC037905.1 | - | 3 | 807 | 5276510 | ||
BT009837.1 | - | 2 | 485 | |||
CR407643.1 | - | 2 | 485 | |||
U79115.1 | - | 3 | 585 | |||
U84388.1 | - | 3 | 522 |