Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261834.1 | NP_001248763.1 | 10, 11 | 1902 | ||
NM_004749.3 | NP_004740.2 | 10, 11 | 1946 | |||
NM_030900.3 | NP_112162.1 | 8, 9 | 1616 | |||
NM_199122.2 | NP_954573.1 | 8, 9 | 1595 | |||
GenBank mRNA | AB023165.1 | - | 8, 9 | 1587 | ||
AF011792.1 | - | 8, 9 | 1280 | |||
AK001281.1 | - | 8, 9 | 1564 | |||
AK022575.1 | - | 10, 11 | 1882 | |||
AK092132.1 | - | 8, 9 | 3080 | |||
AK222943.1 | - | 8, 9 | 1552 | |||
AK298155.1 | - | 8, 9 | 1295 | |||
AK300489.1 | - | 10, 11 | 1861 | |||
AL833840.1 | - | 10, 11 | 1915 | |||
BC002732.2 | - | 8, 9 | 1385 | |||
BC014918.1 | - | 10, 11 | 1876 | 3906670 | ||
BC017235.2 | - | 10, 11 | 1873 | 3538504 | ||
BM150942.1 | - | 1, 2 | 342 |