Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN1 SIRNA Assays ›
Gene Symbol:
NRXN1
Gene Name:
neurexin 1
Gene Aliases:
Hs.22998, PTHSL2, SCZD17
Chromosome Location:
Chr.2: 49918503 - 51032536 on Build GRCh38
UniGene ID:
Hs.637685
Species:
Human
siRNA ID
HSS145132
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135659.1
NP_001129131.1
23, 24
5807
NM_001320156.1
NP_001307085.1
2, 3
318
NM_004801.4
NP_004792.1
21, 22
5597
NM_138735.2
NP_620072.1
5, 6
1822
XM_011533167.2
XP_011531469.1
22, 23
5708
XM_011533171.2
XP_011531473.1
22, 23
5687
XM_011533172.2
XP_011531474.1
21, 22
5681
XM_011533174.2
XP_011531476.1
21, 22
5663
XM_011533175.2
XP_011531477.1
20, 21
5651
XM_011533177.2
XP_011531479.1
21, 22
5618
XM_011533178.2
XP_011531480.1
19, 20
5618
XM_011533183.1
XP_011531485.1
16, 17
3390
XM_017005303.1
XP_016860792.1
23, 24
5735
XM_017005304.1
XP_016860793.1
23, 24
5732
XM_017005306.1
XP_016860795.1
22, 23
5723
XM_017005307.1
XP_016860796.1
22, 23
5717
XM_017005308.1
XP_016860797.1
23, 24
5714
XM_017005309.1
XP_016860798.1
22, 23
5708
XM_017005310.1
XP_016860799.1
21, 22
5705
XM_017005311.1
XP_016860800.1
22, 23
5690
XM_017005312.1
XP_016860801.1
22, 23
5684
XM_017005314.1
XP_016860803.1
20, 21
5675
XM_017005316.1
XP_016860805.1
21, 22
5672
XM_017005318.1
XP_016860807.1
21, 22
5663
XM_017005319.1
XP_016860808.1
21, 22
5660
XM_017005321.1
XP_016860810.1
22, 23
5645
XM_017005323.1
XP_016860812.1
18, 19
5621
XM_017005324.1
XP_016860813.1
20, 21
5591
XM_017005326.1
XP_016860815.1
20, 21
5579
XM_017005328.1
XP_016860817.1
19, 20
5546
XM_017005332.1
XP_016860821.1
16, 17
5486
XM_017005334.1
XP_016860823.1
19, 20
3556
XM_017005335.1
XP_016860824.1
6, 7
6902
GenBank mRNA
AB035356.1
-
20, 21
4312
AF064842.1
-
5, 6
1821
AK295773.1
-
2, 3
212
BC125179.1
-
6, 7
878
BC125180.1
-
6, 7
1241
DC324848.1
-
2, 3
318
DC333548.1
-
2, 3
281
EF539882.1
-
23, 24
5670
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
chemical synaptic transmission
neurotransmitter secretion
axon guidance
synapse assembly
learning
adult behavior
social behavior
vocal learning
neuromuscular process controlling balance
positive regulation of synapse assembly
positive regulation of synaptic transmission, glutamatergic
prepulse inhibition
regulation of insulin secretion involved in cellular response to glucose stimulus
vocalization behavior
positive regulation of synapse maturation
postsynaptic membrane assembly
gephyrin clustering involved in postsynaptic density assembly
neuroligin clustering involved in postsynaptic membrane assembly
postsynaptic density protein 95 clustering
positive regulation of excitatory postsynaptic potential
regulation of grooming behavior
receptor activity
calcium channel regulator activity
calcium ion binding
protein binding
acetylcholine receptor binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0