Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOMER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004839.3 | NP_004830.2 | 5 | 630 | ||
NM_199330.2 | NP_955362.1 | 5 | 663 | |||
XM_005272448.4 | XP_005272505.1 | 5 | 1098 | |||
XM_005272449.3 | XP_005272506.1 | 5 | 510 | |||
XM_006720775.3 | XP_006720838.1 | 5 | 982 | |||
XM_006720776.3 | XP_006720839.1 | 5 | 566 | |||
XM_011522231.2 | XP_011520533.1 | 5 | 982 | |||
XM_011522232.2 | XP_011520534.1 | 5 | 982 | |||
XM_011522233.2 | XP_011520535.1 | 5 | 513 | |||
XM_011522234.2 | XP_011520536.1 | 5 | 780 | |||
GenBank mRNA | AF081530.1 | - | 5 | 504 | ||
AK292772.1 | - | 5 | 596 | |||
BC012109.1 | - | 5 | 627 | 4636423 | ||
Y19025.1 | - | 4 | 441 | |||
Y19026.1 | - | 4 | 474 | |||
Y19027.1 | - | 4 | 441 | |||
Y19028.1 | - | 4 | 474 |