Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321170.1 | NP_001308099.1 | 6, 7 | 462 | ||
NM_138501.5 | NP_612510.1 | 5, 6 | 388 | |||
NR_038103.1 | 5, 6 | 420 | ||||
NR_038104.1 | 5, 6 | 453 | ||||
XM_006722945.1 | XP_006723008.1 | 6, 7 | 506 | |||
XM_011528442.2 | XP_011526744.1 | 5, 6 | 336 | |||
GenBank mRNA | AF038958.1 | - | 5, 6 | 334 | ||
AF222742.1 | - | 5, 6 | 309 | |||
AK001416.1 | - | 5, 6 | 352 | |||
AK094087.1 | - | 6, 7 | 429 | |||
AK299134.1 | - | 6, 7 | 426 | |||
AK315412.1 | - | 5, 6 | 352 | |||
BC000174.2 | - | 4, 5 | 248 | |||
BC002425.1 | - | 5, 6 | 320 | 3163592 | ||
BC005952.1 | - | 5, 6 | 331 | 4249841 | ||
BC007801.2 | - | 5, 6 | 338 | 4128702 | ||
BC013881.2 | - | 5, 6 | 336 | 3928698 | ||
BG705196.1 | - | 5, 6 | 391 | |||
BT007179.1 | - | 4, 5 | 258 | |||
CR456892.1 | - | 4, 5 | 258 |