Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270531.1 | NP_001257460.1 | 4, 5 | 346 | ||
NM_001270532.1 | NP_001257461.1 | 4, 5 | 478 | |||
NM_001270533.1 | NP_001257462.1 | 4, 5 | 346 | |||
NM_004906.4 | NP_004897.2 | 4, 5 | 478 | |||
NM_152857.2 | NP_690596.1 | 4, 5 | 346 | |||
NM_152858.2 | NP_690597.1 | 4, 5 | 478 | |||
XM_017011514.1 | XP_016867003.1 | 4, 5 | 316 | |||
XM_017011515.1 | XP_016867004.1 | 2, 3 | 578 | |||
XM_017011516.1 | XP_016867005.1 | 4, 5 | 316 | |||
GenBank mRNA | AF374416.1 | - | 4, 5 | 359 | ||
AJ276706.1 | - | 3, 4 | 126 | |||
AK127822.1 | - | 4, 5 | 357 | |||
BC000383.2 | - | 4, 5 | 337 | |||
BC004432.1 | - | 4, 5 | 337 | |||
BC069192.1 | - | 4, 5 | 314 | 4653831 | ||
BE906617.1 | - | 4, 5 | 463 | |||
BG702882.1 | - | 4, 5 | 350 | |||
BQ948913.1 | - | 4, 5 | 284 | |||
BX387163.2 | - | 4, 5 | 328 | |||
CN284477.1 | - | 4, 5 | 308 | |||
D14661.1 | - | 4, 5 | 258 | |||
DA138013.1 | - | 4, 5 | 478 | |||
DA561768.1 | - | 4, 5 | 352 |