Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEC16A SIRNA Assays ›
Gene Symbol:
SEC16A
Gene Name:
SEC16 homolog A, endoplasmic reticulum export factor
Gene Aliases:
KIAA0310, SEC16L, p250
Chromosome Location:
Chr.9: 136440096 - 136484552 on Build GRCh38
UniGene ID:
Hs.522500
Species:
Human
siRNA ID
HSS145371
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEC16A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001276418.1
NP_001263347.1
16, 17
5722
NM_014866.1
NP_055681.1
17, 18
5712
XM_005266134.2
XP_005266191.1
16, 17
5575
XM_011519246.2
XP_011517548.1
17, 18
5721
XM_011519247.1
XP_011517549.1
16, 17
5578
XM_011519248.2
XP_011517550.1
16, 17
5552
XM_011519249.1
XP_011517551.1
16, 17
5575
XM_011519250.2
XP_011517552.1
16, 17
5575
XM_011519251.1
XP_011517553.1
16, 17
5575
XM_011519252.2
XP_011517554.1
16, 17
5575
XM_011519253.1
XP_011517555.1
16, 17
5575
XM_011519254.1
XP_011517556.1
16, 17
5575
XM_011519255.2
XP_011517557.1
16, 17
5575
XM_011519256.1
XP_011517558.1
16, 17
5575
XM_011519257.1
XP_011517559.1
16, 17
5575
XM_011519259.1
XP_011517561.1
16, 17
5575
XM_011519260.2
XP_011517562.1
16, 17
5559
XM_011519261.1
XP_011517563.1
13, 14
5128
XM_011519262.1
XP_011517564.1
17, 18
4864
XM_011519263.2
XP_011517565.1
17, 18
4864
XM_011519264.2
XP_011517566.1
16, 17
5575
XM_017015363.1
XP_016870852.1
16, 17
5575
GenBank mRNA
AB002308.4
-
17, 18
5712
AB593115.1
-
16, 17
5722
BC008332.1
-
6, 7
585
BC028183.1
-
12, 13
1579
BC125018.1
-
15, 16
4284
BC125019.1
-
16, 17
3573
DQ903855.1
-
15, 16
4869
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
endoplasmic reticulum organization
protein transport
substantia nigra development
COPII vesicle coating
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0