Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS3ST3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006041.2 | NP_006032.1 | 1 | 591 | ||
NR_130138.1 | 1 | 591 | ||||
GenBank mRNA | AF105377.1 | - | 1 | 452 | ||
AK023723.1 | - | 1 | 558 | |||
BC063301.1 | - | 1 | 508 | 30343352 | ||
BC069664.1 | - | 1 | 392 | 7262483 | ||
BC069725.1 | - | 1 | 391 | 7262495 |