Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS3ST3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006042.2 | NP_006033.1 | 1 | 915 | ||
XM_017025480.1 | XP_016880969.1 | 1 | 915 | |||
GenBank mRNA | AF105376.1 | - | 1 | 900 | ||
AK292047.1 | - | 1 | 914 | |||
AY358838.1 | - | 1 | 874 | |||
BC044647.1 | - | 1 | 265 | 5478042 |