Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMG20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006339.2 | NP_006330.2 | 8 | 684 | ||
XM_017026144.1 | XP_016881633.1 | 8 | 688 | |||
GenBank mRNA | AF072165.1 | - | 5 | 590 | ||
AF072836.1 | - | 5 | 328 | |||
AF146223.1 | - | 7 | 626 | |||
AF288679.1 | - | 7 | 616 | |||
AF318366.1 | - | 6 | 1506 | |||
AF331191.1 | - | 7 | 616 | |||
AK090733.1 | - | 8 | 578 | |||
AK129638.1 | - | 3 | 753 | |||
AK289500.1 | - | 8 | 672 | |||
AK314201.1 | - | 8 | 658 | |||
AL355691.1 | - | 6 | 582 | |||
AL355703.1 | - | 7 | 628 | |||
AL355709.1 | - | 7 | 626 | |||
BC002552.2 | - | 8 | 674 | 3138919 | ||
BC003505.1 | - | 8 | 672 | 3609369 | ||
BC004408.1 | - | 7 | 622 | 3638942 | ||
BC021585.1 | - | 6 | 586 | 4873122 | ||
CR456754.1 | - | 5 | 310 |