Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016941.3 | NP_058637.1 | 1 | 100 | ||
NM_203486.2 | NP_982353.1 | 1 | 100 | |||
GenBank mRNA | AK075302.1 | - | 1 | 70 | ||
AK308460.1 | - | 1 | 122 | |||
BC000218.2 | - | 1 | 49 | 3508262 | ||
DB035825.1 | - | 1 | 100 |