Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256067.1 | NP_001242996.1 | 11 | 1088 | ||
NM_001256068.1 | NP_001242997.1 | 9 | 920 | |||
NM_006647.1 | NP_006638.1 | 11 | 1088 | |||
XM_011518158.2 | XP_011516460.1 | 10 | 1000 | |||
XM_011518159.1 | XP_011516461.1 | 11 | 1106 | |||
XM_011518160.1 | XP_011516462.1 | 12 | 1282 | |||
XM_017014220.1 | XP_016869709.1 | 9 | 1053 | |||
GenBank mRNA | AB095031.1 | - | 11 | 1068 | ||
AF039697.1 | - | 4 | 223 | |||
AK098341.1 | - | 9 | 1013 | |||
AY255769.1 | - | 11 | 953 | |||
AY927790.1 | - | 11 | 991 | |||
BC041594.1 | - | 11 | 1088 | 5442630 | ||
BC110840.1 | - | 11 | 1072 | 5759212 |