Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193270.2 | NP_001180199.1 | 8, 9 | 999 | ||
NM_001282174.1 | NP_001269103.1 | 7, 8 | 931 | |||
NM_006800.3 | NP_006791.2 | 7, 8 | 984 | |||
NM_078628.1 | NP_523352.1 | 8, 9 | 1014 | |||
NM_078629.3 | NP_523353.2 | 8, 9 | 1014 | |||
XM_005274440.2 | XP_005274497.1 | 8, 9 | 891 | |||
XM_011545442.2 | XP_011543744.1 | 8, 9 | 968 | |||
GenBank mRNA | AF117065.1 | - | 8, 9 | 1014 | ||
AK000793.1 | - | 2, 3 | 190 | |||
AK025642.1 | - | 7, 8 | 958 | |||
AK289780.1 | - | 8, 9 | 936 | |||
AK294255.1 | - | 7, 8 | 801 | |||
AK300814.1 | - | 8, 9 | 972 | |||
AL050178.1 | - | 1, 2 | 63 | |||
AL713667.1 | - | 1, 2 | 63 | |||
BC031210.1 | - | 8, 9 | 947 | 5313214 |