Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006830.3 | NP_006821.1 | 2 | 208 | ||
GenBank mRNA | AK312053.1 | - | 2 | 188 | ||
BC000462.2 | - | 2 | 156 | 2822699 | ||
BC003594.1 | - | 2 | 156 | 2822699 | ||
D55636.1 | - | 2 | 170 | |||
DW465790.1 | - | 2 | 213 |