Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281741.1 | NP_001268670.1 | 3, 4 | 410 | ||
NM_007019.3 | NP_008950.1 | 4, 5 | 551 | |||
NM_181799.2 | NP_861515.1 | 4, 5 | 497 | |||
NM_181800.2 | NP_861516.1 | 3, 4 | 464 | |||
NM_181801.3 | NP_861517.1 | 4, 5 | 588 | |||
NR_104036.1 | 2, 3 | 259 | ||||
NR_104037.1 | 3, 4 | 354 | ||||
GenBank mRNA | AK311949.1 | - | 4, 5 | 518 | ||
BC007656.2 | - | 4, 5 | 474 | 3345575 | ||
BC016292.1 | - | 4, 5 | 480 | 3910286 | ||
BC028161.1 | - | 3, 4 | 1090 | |||
BC050736.1 | - | 4, 5 | 501 | 6503915 | ||
BI858659.1 | - | 4, 5 | 588 | |||
BM556795.1 | - | 4, 5 | 415 | |||
BT007300.1 | - | 4, 5 | 431 | |||
BT020165.1 | - | 4, 5 | 431 | |||
BT020166.1 | - | 4, 5 | 431 | |||
BU169315.1 | - | 4, 5 | 438 | |||
BU171488.1 | - | 3, 4 | 370 | |||
BU844974.1 | - | 3, 4 | 366 | |||
CB143441.1 | - | 2, 3 | 177 | |||
CK904708.1 | - | 3, 4 | 376 | |||
CN354815.1 | - | 3, 4 | 289 | |||
CR456988.1 | - | 4, 5 | 431 | |||
U73379.1 | - | 4, 5 | 471 |