Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007213.2 | NP_009144.1 | 1 | 92 | ||
GenBank mRNA | AJ005896.1 | - | 1 | 50 | ||
AK023433.1 | - | 1 | 49 | |||
AK310550.1 | - | 1 | 47 | |||
AK315374.1 | - | 1 | 49 | |||
BC021213.2 | - | 1 | 36 | 4297902 | ||
CB155577.1 | - | 1 | 92 | |||
CR457023.1 | - | 1 | 34 |