Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042540.1 | NP_001036005.1 | 3 | 248 | ||
NM_001308036.1 | NP_001294965.1 | 3 | 745 | |||
NM_007362.3 | NP_031388.2 | 3 | 407 | |||
XM_011512556.2 | XP_011510858.1 | 3 | 464 | |||
XM_011512557.2 | XP_011510859.1 | 3 | 512 | |||
XM_011512558.2 | XP_011510860.1 | 3 | 354 | |||
GenBank mRNA | AK093216.1 | - | 3 | 457 | ||
AK296144.1 | - | 3 | 371 | |||
AK297506.1 | - | 3 | 710 | |||
AK308192.1 | - | 3 | 411 | |||
AK315903.1 | - | 3 | 381 | |||
AK316601.1 | - | 3 | 374 | |||
AY644767.1 | - | 3 | 331 | |||
BC001255.1 | - | 3 | 343 | 3458927 | ||
BP248419.1 | - | 3 | 407 | |||
BT006842.1 | - | 3 | 317 | |||
BX537372.1 | - | 2 | 1975 | |||
BX647994.1 | - | 3 | 392 | |||
CN421262.1 | - | 3 | 193 | |||
D59253.1 | - | 3 | 353 | |||
X84157.1 | - | 3 | 346 |