Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014981.1 | NP_055796.1 | 38, 39 | 5553 | ||
XM_011512559.2 | XP_011510861.1 | 39, 40 | 5674 | |||
XM_017005922.1 | XP_016861411.1 | 29, 30 | 4818 | |||
GenBank mRNA | AB023217.3 | - | 38, 39 | 5553 |