Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277285.1 | NP_001264214.1 | 9 | 1132 | ||
XM_011542691.2 | XP_011540993.1 | 10 | 1807 | |||
XM_011542692.2 | XP_011540994.1 | 10 | 1807 | |||
XM_011542694.2 | XP_011540996.1 | 10 | 1810 | |||
XM_011542695.2 | XP_011540997.1 | 9 | 1338 | |||
XM_017017393.1 | XP_016872882.1 | 9 | 1825 | |||
XM_017017394.1 | XP_016872883.1 | 10 | 1810 | |||
XM_017017395.1 | XP_016872884.1 | 10 | 1809 | |||
XM_017017396.1 | XP_016872885.1 | 3 | 1149 | |||
GenBank mRNA | AK097578.1 | - | 6 | 862 |