Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276252.1 | NP_001263181.1 | 3 | 605 | ||
NM_015023.4 | NP_055838.2 | 3 | 605 | |||
XM_011541057.1 | XP_011539359.1 | 3 | 283 | |||
XM_011541058.2 | XP_011539360.1 | 3 | 550 | |||
XM_017000730.1 | XP_016856219.1 | 3 | 550 | |||
GenBank mRNA | AK001734.1 | - | 2 | 77 | ||
AK315564.1 | - | 3 | 489 | |||
BC032523.1 | - | 3 | 468 | 5500384 |