Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESYT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184796.1 | NP_001171725.1 | 10 | 1312 | ||
NM_015292.2 | NP_056107.1 | 10 | 1312 | |||
GenBank mRNA | AB018290.1 | - | 10 | 1069 | ||
AK022790.1 | - | 10 | 1263 | |||
AK025463.1 | - | 6 | 517 | |||
AK128730.1 | - | 9 | 1116 | |||
AK225035.1 | - | 10 | 1247 | |||
AK290337.1 | - | 10 | 1247 | |||
AL050134.1 | - | 2 | 60 | |||
BC004998.1 | - | 10 | 1244 | 2958094 | ||
DQ993200.1 | - | 10 | 1194 |