Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2SURP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080415.1 | NP_001073884.1 | 6 | 616 | ||
NM_001320219.1 | NP_001307148.1 | 6 | 616 | |||
NM_001320220.1 | NP_001307149.1 | 6 | 616 | |||
NM_001320222.1 | NP_001307151.1 | 6 | 616 | |||
XM_005247248.4 | XP_005247305.1 | 6 | 622 | |||
XM_017006037.1 | XP_016861526.1 | 3 | 295 | |||
XM_017006038.1 | XP_016861527.1 | 6 | 555 | |||
XM_017006039.1 | XP_016861528.1 | 1 | 66 | |||
GenBank mRNA | AB002330.1 | - | 6 | 515 | ||
AK296440.1 | - | 6 | 536 | |||
BC006474.1 | - | 6 | 515 | |||
BC016323.1 | - | 6 | 533 | |||
BC105604.1 | - | 6 | 548 | |||
BC111692.1 | - | 6 | 987 | 40028600 |