Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDNEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143775.1 | NP_001137247.1 | 5 | 835 | ||
NM_015343.4 | NP_056158.2 | 6 | 902 | |||
GenBank mRNA | AJ011916.1 | - | 5 | 485 | ||
AY364239.1 | - | 5 | 455 | |||
BC009295.2 | - | 6 | 901 | 4123279 |