Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206739.1 | NP_001193668.1 | 8, 9 | 729 | ||
NM_014288.4 | NP_055103.3 | 7, 8 | 612 | |||
XM_017000855.1 | XP_016856344.1 | 6, 7 | 625 | |||
GenBank mRNA | AA873717.1 | - | 2, 3 | 404 | ||
AF175306.1 | - | 7, 8 | 503 | |||
AK312944.1 | - | 7, 8 | 564 | |||
BC009929.2 | - | 7, 8 | 519 | 3505579 | ||
BC014385.1 | - | 8, 9 | 654 | 4064625 |