Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007352.3 | NP_031378.1 | 2 | 229 | ||
GenBank mRNA | AK092382.1 | - | 2 | 102 | ||
AK315798.1 | - | 2 | 98 | |||
BC005216.1 | - | 2 | 103 | 3949903 | ||
BI760781.1 | - | 2 | 228 | |||
BX499439.1 | - | 2 | 198 | |||
M16630.1 | - | 2 | 74 | |||
M18692.1 | - | 2 | 101 |
Gene Symbol: CELA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308510.1 | - | 2 | 95 | ||
AK308518.1 | - | 2 | 102 |