Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1549L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012194.2 | NP_036326.2 | 10 | 3765 | ||
XM_005252847.3 | XP_005252904.2 | 12 | 3859 | |||
XM_005252848.3 | XP_005252905.1 | 10 | 3783 | |||
XM_011519969.2 | XP_011518271.1 | 10 | 3783 | |||
XM_011519970.2 | XP_011518272.1 | 11 | 2425 | |||
XM_017017484.1 | XP_016872973.1 | 10 | 3783 | |||
XM_017017485.1 | XP_016872974.1 | 10 | 3783 | |||
XM_017017486.1 | XP_016872975.1 | 11 | 2425 | |||
GenBank mRNA | AK124395.1 | - | 10 | 3783 | ||
U10991.1 | - | 10 | 3172 |