Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242374.1 | NP_001229303.1 | 2 | 771 | ||
NM_001242375.1 | NP_001229304.1 | 2 | 356 | |||
NM_015391.3 | NP_056206.1 | 2 | 137 | |||
GenBank mRNA | AF086169.1 | - | 1 | 389 | ||
AK001285.1 | - | 2 | 768 | |||
AK026550.1 | - | 2 | 91 | |||
AK290761.1 | - | 2 | 367 | |||
AL050051.1 | - | 2 | 356 | |||
AV682398.1 | - | 2 | 102 | |||
BC005398.1 | - | 2 | 103 | 3828623 | ||
BU852706.1 | - | 2 | 117 | |||
BX537696.1 | - | 3 | 557 | |||
CN258858.1 | - | 2 | 337 |