Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSKU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258210.1 | NP_001245139.1 | 2 | 614 | ||
NM_001318477.1 | NP_001305406.1 | 2 | 1511 | |||
NM_001318478.1 | NP_001305407.1 | 2 | 711 | |||
NM_001318479.1 | NP_001305408.1 | 2 | 522 | |||
NM_015516.3 | NP_056331.2 | 2 | 657 | |||
GenBank mRNA | AF191019.1 | - | 1 | 498 | ||
AK075476.1 | - | 2 | 569 | |||
AK091475.1 | - | 3 | 474 | |||
AK126536.1 | - | 2 | 1511 | |||
AK313405.1 | - | 2 | 570 | |||
AL110276.1 | - | 1 | 153 | |||
AY358317.1 | - | 2 | 548 | |||
BC020975.1 | - | 2 | 535 | 3848274 | ||
BT007440.1 | - | 1 | 485 | |||
DA879213.1 | - | 2 | 522 |