Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IBTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300906.1 | NP_001287835.1 | 14 | 2885 | ||
NM_015525.3 | NP_056340.2 | 14 | 2885 | |||
XM_006715453.2 | XP_006715516.1 | 14 | 3425 | |||
XM_006715454.2 | XP_006715517.1 | 14 | 3425 | |||
GenBank mRNA | AB037838.2 | - | 13 | 2609 | ||
BC113696.1 | - | 13 | 2414 | 8322748 | ||
BC113698.1 | - | 13 | 2414 | 8322750 | ||
BC143736.1 | - | 12 | 1811 | 9052250 | ||
BC143745.1 | - | 13 | 2415 | 9052259 | ||
BC143748.1 | - | 12 | 1811 | |||
DQ005633.1 | - | 14 | 2885 | |||
DQ005634.1 | - | 14 | 2885 |