Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015626.8 | NP_056441.6 | 4 | 815 | ||
NM_134265.2 | NP_599027.1 | 2 | 377 | |||
XM_005257963.3 | XP_005258020.1 | 3 | 612 | |||
XM_017024433.1 | XP_016879922.1 | 4 | 815 | |||
XM_017024434.1 | XP_016879923.1 | 5 | 903 | |||
XM_017024435.1 | XP_016879924.1 | 5 | 914 | |||
XM_017024436.1 | XP_016879925.1 | 3 | 646 | |||
GenBank mRNA | AF069313.2 | - | 4 | 764 | ||
AF072880.1 | - | 4 | 590 | |||
AF106683.1 | - | 4 | 632 | |||
AF112205.1 | - | 4 | 754 | |||
AF240696.1 | - | 2 | 326 | |||
AK057082.1 | - | 2 | 1222 | |||
AK074917.1 | - | 4 | 781 | |||
AK222576.1 | - | 4 | 781 | |||
AK294516.1 | - | 4 | 688 | |||
AK300262.1 | - | 4 | 781 | |||
AK307114.1 | - | 2 | 354 | |||
AL110243.1 | - | 4 | 815 | |||
BC021110.1 | - | 4 | 762 | 4865448 | ||
BC048007.1 | - | 4 | 781 |