Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080453.2 | NP_001073922.2 | 35 | 4957 | ||
XM_011515260.1 | XP_011513562.1 | 35 | 4957 | |||
XM_017011959.1 | XP_016867448.1 | 35 | 4957 | |||
XM_017011960.1 | XP_016867449.1 | 35 | 4957 | |||
GenBank mRNA | AB037861.1 | - | 17 | 2417 | ||
AL050110.1 | - | 2 | 207 | |||
AY358482.1 | - | 10 | 1418 | |||
BC004286.1 | - | 16 | 2130 | |||
BC013367.2 | - | 5 | 713 | |||
BC018777.1 | - | 3 | 380 | |||
BC069262.1 | - | 7 | 987 |