Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012166.2 | NP_036298.2 | 3 | 1217 | ||
XM_005251439.4 | XP_005251496.1 | 3 | 1306 | |||
XM_017014618.1 | XP_016870107.1 | 3 | 1423 | |||
XM_017014619.1 | XP_016870108.1 | 3 | 1534 | |||
GenBank mRNA | AB209610.1 | - | 2 | 805 | ||
AF176705.1 | - | 3 | 1216 | |||
BC125125.1 | - | 3 | 1296 | 40119019 | ||
BC140785.1 | - | 2 | 1188 | 9021653 | ||
BC171785.1 | - | 2 | 1188 | 9054439 |