Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CECR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015352.1 | 4 | 1872 | |||
GenBank mRNA | AK098045.1 | - | 4 | 1203 | ||
AY026052.1 | - | 4 | 1872 |