Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKIRAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001349.2 | NP_001001349.1 | 3 | 308 | ||
NM_001144927.1 | NP_001138399.1 | 3 | 412 | |||
NM_017595.5 | NP_060065.2 | 3 | 393 | |||
XM_005257251.1 | XP_005257308.1 | 3 | 420 | |||
GenBank mRNA | AF229840.1 | - | 2 | 243 | ||
AK027265.1 | - | 3 | 384 | |||
AK054571.1 | - | 3 | 420 | |||
AL137682.1 | - | 1 | 2133 | |||
BC007316.2 | - | 3 | 345 | |||
BC007450.2 | - | 3 | 301 | 3028708 | ||
BC063498.1 | - | 3 | 864 | 5248950 | ||
BG717103.1 | - | 3 | 414 | |||
DA607452.1 | - | 3 | 391 |