Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014050.3 | NP_054769.1 | 3 | 278 | ||
NM_172177.3 | NP_751917.1 | 3 | 281 | |||
NR_038159.1 | 4 | 301 | ||||
NR_038160.1 | 4 | 298 | ||||
NR_038161.1 | 4 | 383 | ||||
GenBank mRNA | AF078860.1 | - | 3 | 260 | ||
AF135160.1 | - | 3 | 264 | |||
AF151038.1 | - | 3 | 252 | |||
AK000285.1 | - | 3 | 254 | |||
AL136659.1 | - | 3 | 255 | |||
BC040240.2 | - | 2 | 217 | 3445213 | ||
BM147739.1 | - | 3 | 279 | |||
BM726733.1 | - | 4 | 256 | |||
BU561400.1 | - | 277 | ||||
CD385500.1 | - | 3 | 274 | |||
CR533495.1 | - | 2 | 119 | |||
CR749344.1 | - | 4 | 355 | |||
DB195199.1 | - | 4 | 359 |