Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014050.3 | NP_054769.1 | 5, 6 | 532 | ||
NM_172177.3 | NP_751917.1 | 5, 6 | 535 | |||
NR_038159.1 | 6, 7 | 555 | ||||
NR_038160.1 | 6, 7 | 552 | ||||
NR_038161.1 | 6, 7 | 637 | ||||
GenBank mRNA | AF078860.1 | - | 5, 6 | 514 | ||
AF135160.1 | - | 5, 6 | 518 | |||
AF151038.1 | - | 5, 6 | 506 | |||
AK000285.1 | - | 5, 6 | 508 | |||
AL136659.1 | - | 5, 6 | 509 | |||
BC040240.2 | - | 4, 5 | 471 | 3445213 | ||
BM726733.1 | - | 6, 7 | 510 | |||
BU561400.1 | - | 531 | ||||
CD385500.1 | - | 5, 6 | 528 | |||
CR533495.1 | - | 4, 5 | 373 | |||
CR749344.1 | - | 6, 7 | 609 |