Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324458.1 | NP_001311387.1 | 6, 7 | 505 | ||
NM_001324459.1 | NP_001311388.1 | 6, 7 | 533 | |||
NM_013292.4 | NP_037424.2 | 4, 5 | 368 | |||
GenBank mRNA | AF363061.1 | - | 4, 5 | 341 | ||
AK314082.1 | - | 4, 5 | 368 | |||
BC012571.1 | - | 4, 5 | 346 | 4245958 | ||
BI828249.1 | - | 6, 7 | 533 | |||
BX649054.1 | - | 4, 5 | 336 | |||
CR456963.1 | - | 3, 4 | 287 | |||
M21812.1 | - | 4, 5 | 348 | |||
X57543.1 | - | 4, 5 | 348 |