Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC35C2 SIRNA Assays ›
Gene Symbol:
SLC35C2
Gene Name:
solute carrier family 35 member C2
Gene Aliases:
BA394O2.1, C20orf5, CGI-15, OVCOV1
Chromosome Location:
Chr.20: 46349528 - 46364461 on Build GRCh38
UniGene ID:
Hs.593344
Species:
Human
siRNA ID
HSS147226
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC35C2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281457.1
NP_001268386.1
2
410
NM_001281458.1
NP_001268387.1
3
278
NM_001281459.1
NP_001268388.1
2
410
NM_001281460.1
NP_001268389.1
3
742
NM_015945.11
NP_057029.8
2
410
NM_173073.3
NP_775096.1
2
410
NM_173179.3
NP_775271.1
2
605
XM_011528831.1
XP_011527133.1
2
588
XM_011528832.1
XP_011527134.1
2
882
XM_011528833.1
XP_011527135.1
2
406
XM_011528834.1
XP_011527136.1
2
410
XM_011528835.1
XP_011527137.1
2
410
XM_011528836.1
XP_011527138.1
2
562
XM_011528837.1
XP_011527139.1
2
396
XM_011528838.1
XP_011527140.1
1
517
XM_017027856.1
XP_016883345.1
2
1305
XM_017027857.1
XP_016883346.1
2
833
XM_017027858.1
XP_016883347.1
2
743
XM_017027859.1
XP_016883348.1
2
1110
XM_017027860.1
XP_016883349.1
3
550
XM_017027861.1
XP_016883350.1
2
572
XM_017027862.1
XP_016883351.1
2
410
GenBank mRNA
AF132949.1
-
2
347
AF455052.1
-
2
365
AK222928.1
-
2
350
AK300207.1
-
2
364
AK300798.1
-
2
342
AK301022.1
-
3
220
AK315726.1
-
2
572
BC014191.2
-
2
354
4761663
BC021138.1
-
2
548
4870715
BC025277.1
-
2
606
4865888
BI831689.1
-
3
354
BM846744.1
-
2
376
DA820126.1
-
2
410
DB023133.1
-
3
493
DB246755.1
-
2
365
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
negative regulation of gene expression
UDP-glucose transport
protein O-linked fucosylation
positive regulation of Notch signaling pathway
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0