Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137602.2 | NP_001131074.1 | 4 | 730 | ||
NM_001301833.1 | NP_001288762.1 | 6 | 510 | |||
NM_001301852.1 | NP_001288781.1 | 5 | 432 | |||
NM_015955.3 | NP_057039.1 | 5 | 799 | |||
NR_126032.1 | 5 | 799 | ||||
NR_126034.1 | 5 | 432 | ||||
XM_011532892.2 | XP_011531194.1 | 6 | 447 | |||
XM_011532896.2 | XP_011531198.1 | 4 | 325 | |||
XM_017004255.1 | XP_016859744.1 | 6 | 420 | |||
GenBank mRNA | AB041018.1 | - | 5 | 799 | ||
AB041019.1 | - | 5 | 448 | |||
AF132961.1 | - | 5 | 497 | |||
AF363446.1 | - | 5 | 716 | |||
AF529368.1 | - | 5 | 382 | |||
AK057760.1 | - | 5 | 421 | |||
AK290753.1 | - | 6 | 466 | |||
AK297128.1 | - | 4 | 441 | |||
BC018733.1 | - | 5 | 764 | 4851517 | ||
BC070046.1 | - | 6 | 539 | 5273070 | ||
BC094681.1 | - | 5 | 403 | 6205399 | ||
BG033287.1 | - | 5 | 381 | |||
BP248979.1 | - | 6 | 510 | |||
CD250945.1 | - | 5 | 496 |